Kaliumbromid ≥99%

Leverandør: Thermo Fisher Scientific
Warning

A16339.0B A16339.0I A16339.22 A16339.30 A16339.36 A16339.A4
A16339.0BEA 1150 NOK
A16339.0B A16339.0I A16339.22 A16339.30 A16339.36 A16339.A4
Kaliumbromid ≥99%
Potassium bromide

Purity: 99+% CAS No.: 7758-02-3 Molecular Formula: KBr Formula Weight: 119.01 Refractive Index: 1.559 Odour: Odourless Boiling Point: 1435° Merck Reference: 14,7618 MDL No.: MFCD00011358

Formel: KBr
Kokepunkt: 1435 °C
Smeltepunkt: 730 °C
Tetthet: 2,75
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011358
CAS-nummer: 7758-02-3
EINECS: 231-830-3
Merck Indeks: 14,7618

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR