Cupferron ≥97%

Leverandør: Thermo Fisher Scientific
Danger

A16551.14
A16551.14EA 480 NOK
A16551.14
Cupferron ≥97%
Cupferron

Purity: 97+% CAS No.: 135-20-6 Molecular Formula: C6H9N3O2 Formula Weight: 155.16 Storage Temperature: 2°C to 8°C Odour: Odourless Merck Reference: 14,2622 UN No.: 2811 MDL No.: MFCD00078422

Formel: C₆H₉N₃O₂
Smeltepunkt: 154 °C (dec.)
MDL nummer: MFCD00078422
CAS-nummer: 135-20-6
EINECS: 205-183-2
FN: 2811
ADR: 6.1,III
Merck Indeks: 14,2622

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR