2-Fluoro-p-toluidine ≥99%

Leverandør: Thermo Fisher Scientific
Danger

A16591.06 A16591.14
A16591.06EA 815 NOK
A16591.06 A16591.14
2-Fluoro-p-toluidine ≥99%
2-Fluoro-p-toluidine

Purity: 99% CAS No.: 452-80-2 Molecular Formula: C7H8FN Formula Weight: 125.15 Flash Point: 79°(174°F) Refractive Index: 1.5330 Boiling Point: 70-71°/7mm Beilstein Reference: 2637578 UN No.: 2810 MDL No.: MFCD00040975

Formel: C₇H₈FN
Kokepunkt: 70…71 °C (7 mmHg)
Smeltepunkt: 3 °C
Tetthet: 1,108
Flash punkt: 79 °C (174 °F)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00040975
CAS-nummer: 452-80-2
FN: 2810
ADR: 6.1,III

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR