2,2':5',2''-Terthiophene ≥99%

Leverandør: Thermo Fisher Scientific

A16748.03 A16748.06 A16748.MD
A16748.03EA 3525 NOK
A16748.03 A16748.06 A16748.MD
2,2':5',2''-Terthiophene ≥99%
2,2':5',2''-Terthiophene

Purity: 99% CAS No.: 1081-34-1 Molecular Formula: C12H8S3 Formula Weight: 248.39 Melting Point: 89-94° Boiling Point: 160°/0.1mm Merck Reference: 14,9174 MDL No.: MFCD00012167

Formel: C₁₂H₈S₃
Kokepunkt: 160 °C (0,1 mmHg)
Smeltepunkt: 89…94 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012167
CAS-nummer: 1081-34-1
Merck Indeks: 14,9174

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR