D(+)-Glukose, vannfri 99%

Leverandør: Thermo Fisher Scientific

A16828.36 A16828.0C A16828.0E
A16828.36EA 440 NOK
A16828.36 A16828.0C A16828.0E
D(+)-Glukose, vannfri 99%
D(+)-Glukose

Purity: 99% CAS No.: 50-99-7 Molecular Formula: C6H12O6 Formula Weight: 180.16 Physical Form: Cryst powder Appearance: White Melting Point: 147-153° Beilstein Reference: 1281608 Merck Reference: 14,4459 Optical Rotation: +53° (c=10 in water, 3h) MDL No.: MFCD00063774

Formel: C₆H₁₂O₆
Smeltepunkt: 147…153 °C
Tetthet: 1,54 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00148912
CAS-nummer: 50-99-7
EINECS: 200-075-1
Merck Indeks: 14,4459
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ec28c7fd4803097405f8eaef3171b3a40d40c5&vid=62627c96e2ae9f2a0cae732d0ddf0a100247298e&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR