3-Nitrophenyl isocyanate ≥97%

Leverandør: Thermo Fisher Scientific
Danger

Synonymer: 1-Isocyanato-3-nitrobenzene, 3-Nitrophenylisocyanate

A16920.06 A16920.14
A16920.06EA 785 NOK
A16920.06 A16920.14
3-Nitrophenyl isocyanate ≥97%
3-Nitrophenyl isocyanate

Purity: 97% CAS No.: 3320-87-4 Molecular Formula: C7H4N2O3 Formula Weight: 164.12 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Odour: Irritating Melting Point: 51-53° Boiling Point: 130-131°/11mm UN No.: 2811 MDL No.: MFCD00007220

Formel: C₇H₄N₂O₃
MW: 164.12 g/mol
Kokepunkt: 130…131 °C (11 mmHg)
Smeltepunkt: 51…53 °C
Flash punkt: >110 °C (230 °F)
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00007220
CAS-nummer: 3320-87-4
EINECS: 222-025-8
FN: 2811
ADR: 6.1 III

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR