4-Methoxystilbene ≥98%

Leverandør: Thermo Scientific

A17328.06
A17328.06EA 1325 NOK
A17328.06
4-Methoxystilbene ≥98%
4-Methoxystilbene

Purity: 98% CAS No.: 1142-15-0 Molecular Formula: C15H14O Formula Weight: 210.27 Melting Point: 136-138° MDL No.: MFCD00017178

Formel: C₁₅H₁₄O
MW: 210,27 g/mol
Smeltepunkt: 136…138 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00017178
CAS-nummer: 1142-15-0
EINECS: 214-530-7

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR