Azure B (azure I)

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: C.I. 52010, N,N,N′-Trimethylthionin, Azure B, Trimethylthionine chloride, 3-(Dimethylamino)-7-(methylamino)phenothiazin-5-ium chloride, Methylene Azur B

A17508.06 A17508.14 A17508.22
A17508.06EA 965 NOK
A17508.06 A17508.14 A17508.22
Azure B (azure I)
Azure B (azure I)

CAS No.: 531-55-5 Molecular Formula: C15H16ClN3S Formula Weight: 305.83 Odour: Odourless Melting Point: ca 205° dec. Beilstein Reference: 3922630 Merck Reference: 14,6058 MDL No.: MFCD00011935

Formel: C₁₅H₁₆ClN₃S
MW: 305.83 g/mol
Smeltepunkt: ∼ 205 °C (dec.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011935
CAS-nummer: 531-55-5
EINECS: 208-511-2

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR