1,3-Dihydroksynaftalen ≥98%

Leverandør: Thermo Fisher Scientific
Warning

A17739.06 A17739.03 A17739.14
A17739.06EA 2450 NOK
A17739.06 A17739.03 A17739.14
1,3-Dihydroksynaftalen ≥98%
1,3-Dihydroksynaftalen

Purity: 98% CAS No.: 132-86-5 Molecular Formula: C10H8O2 Formula Weight: 160.17 Melting Point: 123-126° Beilstein Reference: 2044002 Merck Reference: 14,6396 MDL No.: MFCD00003965

Formel: C₁₀H₈O₂
Kokepunkt: 386 °C (1013 hPa)
Smeltepunkt: 123…126 °C
Tetthet: 1,12 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003965
CAS-nummer: 132-86-5
EINECS: 205-079-7
Merck Indeks: 14,6396

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR