4,4'-Bi-o-toluidin ≥98%

Leverandør: Thermo Fisher Scientific
Danger

A17837.14 A17837.22
A17837.14EA 1200 NOK
A17837.14 A17837.22
4,4'-Bi-o-toluidin ≥98%
4,4'-Bi-o-toluidin

Purity: 98% CAS No.: 119-93-7 Molecular Formula: C14H16N2 Formula Weight: 212.30 Flash Point: 244°(471°F) Melting Point: 128-132° Boiling Point: 300-301° Merck Reference: 14,9514 UN No.: 3077 MDL No.: MFCD00014773

Formel: [-C₆H₃(CH₃)-₄-NH₂]₂
Kokepunkt: 300…301 °C
Smeltepunkt: 128…132 °C
Tetthet: ∼1,234 g/cm³ (20 °C)
Flash punkt: 244 °C (471 °F)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00014773
CAS-nummer: 119-93-7
EINECS: 204-358-0
FN: 3077
ADR: 9,III
Merck Indeks: 14,9514

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR