Wright's stain for microscopical staining

Leverandør: Thermo Fisher Scientific

Synonymer: Eosin Methylene blue solution according to Wright, Wright's eosin methylene blue

A17903.14 A17903.22
A17903.14EA 850 NOK
A17903.14 A17903.22
Wright's stain for microscopical staining
Wright's stain
Tetthet: 0.8 g/cm³ (20 °C)
MDL nummer: MFCD00082143
CAS-nummer: 68988-92-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR