3-Chloro-o-toluidine ≥98%

Leverandør: Thermo Fisher Scientific
Danger

A18221.30
A18221.30EA 2075 NOK
A18221.30
3-Chloro-o-toluidine ≥98%
3-Chloro-o-toluidine

Purity: 98+% CAS No.: 87-60-5 Molecular Formula: C7H8ClN Formula Weight: 141.60 Flash Point: 113°(235°F) Refractive Index: 1.5890 Melting Point: 2° Boiling Point: 245° Beilstein Reference: 471697 UN No.: 3429 MDL No.: MFCD00007766

Formel: C₇H₈ClN
Kokepunkt: 245 °C
Smeltepunkt: 2 °C
Tetthet: 1,188
Flash punkt: 113 °C (235 °F)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007766
CAS-nummer: 87-60-5
EINECS: 201-756-6
FN: 3429
ADR: 6.1,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR