Ethyl red

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: 4-(Diethylamino)azobenzene-2'-carboxylic Acid, Diethyl Red

A18367.09 A18367.18 A18367.30
A18367.09EA 530 NOK
A18367.09 A18367.18 A18367.30
Ethyl red
Ethyl red

CAS No.: 76058-33-8 Molecular Formula: C17H19N3O2 Formula Weight: 297.36 Melting Point: ca 135° dec. Beilstein Reference: 3366056 MDL No.: MFCD00002427

Formel: C₁₇H₁₉N₃O₂
MW: 297.36 g/mol
Smeltepunkt: ∼ 135 °C (dec.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002427
CAS-nummer: 76058-33-8

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR