Penicillin G kaliumsalt

Leverandør: PanReac AppliChem
Warning

Synonymer: 6-(2-Phenylacetamido)penicillanic acid potassium salt, Potassium (2S,5R,6R)-3,3-dimethyl-7-oxo-6-(2-phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, Penicillin G potassium, Potassium [2S-(2α,5α,6β)]-3,3-dimethyl-7-oxo-6-(phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, Benzylpenicillin potassium salt

A1837.0100
A1837.0100EA 2400 NOK
A1837.0100
Penicillin G kaliumsalt
Penicillin G kaliumsalt
Formel: C₁₆H₁₇KN₂O₄S
Smeltepunkt: 214…217 °C
Tetthet: 1,359 g/cm³ (20 °C)
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00036193
CAS-nummer: 113-98-4
EINECS: 204-038-0
Merck Indeks: 12,07225

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR