Spermidine 99

Leverandør: Thermo Fisher Scientific
Danger

Synonymer: N1-(3-Aminopropyl)butane-1,4-diamine, N-(3-Aminopropyl)-1,4-diaminobutane, 4-Azaoctamethylenediamine

A19096.03 A19096.06 A19096.14 A19096.22
A19096.03EA 625 NOK
A19096.03 A19096.06 A19096.14 A19096.22
Spermidine 99
Spermidine

Purity: 99% CAS No.: 124-20-9 Molecular Formula: H2N(CH2)3NH(CH2)4NH2 Formula Weight: 145.25 Flash Point: 110°(230°F) Refractive Index: 1.4790 Melting Point: 22-25° Boiling Point: 128-130°/14mm Beilstein Reference: 1698591 Merck Reference: 14,8742 UN No.: 2735 MDL No.: MFCD00008229

Formel: C₇H₁₉N₃
MW: 145.25 g/mol
Kokepunkt: 128…130 °C (14 mmHg)
Smeltepunkt: 22…25 °C
Tetthet: 0.925
Flash punkt: >110 °C (230 °F)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008229
CAS-nummer: 124-20-9
EINECS: 204-689-0
ADR: 8 III
Merck Indeks: 12,08893

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR