Tetrabutylammonium dihydrogen phosphate

Leverandør: PanReac AppliChem

Synonymer: Tetra-n-butylammonium phosphate monobasic, Tetrabutylammonium phosphate monobasic, Tetra-n-butylammonium dihydrogen phosphate, IPC-TBA-P, Tetra-n-butylammonium phosphate

A1947.0025
A1947.0025EA 5675 NOK
A1947.0025
Tetrabutylammonium dihydrogen phosphate
Tetrabutylammonium dihydrogen phosphate
Formel: C₁₆H₃₈NO₄P
MDL nummer: MFCD00064526
CAS-nummer: 5574-97-0
EINECS: 226-947-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR