1,3-Diphenyl-2-thiourea ≥98%

Leverandør: Thermo Fisher Scientific
Danger

A19644.0E A19644.22 A19644.36
A19644.0EEA 2200 NOK
A19644.0E A19644.22 A19644.36
1,3-Diphenyl-2-thiourea ≥98%
1,3-Diphenyl-2-thiourea

Purity: 98% CAS No.: 102-08-9 Molecular Formula: C13H12N2S Formula Weight: 228.32 Melting Point: 150-154° Beilstein Reference: 644277 Merck Reference: 14,3333 UN No.: 2811 MDL No.: MFCD00004921

Formel: C₁₃H₁₂N₂S
Kokepunkt: 365 °C (1013 hPa)
Smeltepunkt: 150…154 °C
Tetthet: 1,320
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004921
CAS-nummer: 102-08-9
EINECS: 203-004-2
FN: 2811
ADR: 6.1,II
Merck Indeks: 14,3333

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR