Acridine 97%, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Warning


M-Clarity™ quality level = MQ100

A23609-5G A23609-25G
A23609-5GEA 543 NOK
A23609-5G A23609-25G
Acridine 97%, Sigma-Aldrich®
Acridine

Acridine is a small cationic and planar dye that mainly binds to RNA and DNA. These dyes are strongly fluorescent. It resembles the structure of xanthene except that the heteroatom is nitrogen, not oxygen. Acridine is suitable for nucleic acid staining, particularly for cell cycle determination. Acridines are used as fluorescent materials for visualisation of biomolecules, and in laser technologies.

Formel: C₁₃H₉N
Kokepunkt: 346 °C (lit.)
Lagringstemperatur: Room Temperature
MDL nummer: MFCD00005025
CAS-nummer: 260-94-6
EINECS: 205-971-6
FN: 2713
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6fa758dffbcc4d600937cc56d2f11cb80624e845&vid=1671db5d1a6f3467bf2093285463ff09da8212fe&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance (Colour) Light Yellow to Very Dark Yellow and Light Beige to Beige and Light Brown to Brown
Appearance (Form) Powder or Crystals
Infrared spectrum Conforms to Structure
Purity (Titration by HCL04) 96.5 - 103.5%
Purity (HPLC) ≥96.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR