Azure A, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Warning

Synonymer: Azurol a, N,N-Dimethylthionine, Azure A chloride, azurol I, 7 - (dimethyl) - 3-imino-3h-dithizine hydrochloride, 3-amino-7-dimethylaminophenothiazine-5-onium chloride, 3-Amino-7-(dimethylamino)phenothiazin-5-ium chloride, 3-Amino-7-(dimethylamino)phenazathionium Chloride, Methylene Azur A

M-Clarity™ quality level = MQ200

A2918-25G
A2918-25GEA 3870 NOK
A2918-25G
Azure A, Sigma-Aldrich®
Azure A
Formel: C₁₄H₁₄ClN₃S
MW: 291.797 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012112
CAS-nummer: 531-53-3
EINECS: 208-510-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d628de8a8dd6a7ea5fbc184239f4f9330c12215f&vid=df9187f4e13c3629cf11e7793a4b3aec7d481ed5&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR