Yeast extract, molecular biology grade
Leverandør: PanReac AppliChem
A3732.0500
A3732.0500EA 2400 NOK
A3732.0500
Yeast extract, molecular biology grade
Cellekultursupplementer og tilsetninger
Water-soluble part of autolysed yeast. Rich of ammonia and vitamins, especially of water-soluble vitamin B-complexes; suitable for addition to media.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Arter
Kilde