Spectinomycin dihydrochloride pentahydrate ≥95% (by titrimetric analysis), BioChemica®

Leverandør: PanReac AppliChem

Synonymer: (2R,4aR,5aR,6S,7S,8R,9S,9aR,10aS)-4a,7,9-Trihydroxy-2-methyl-6,8-bis(methylamino)decahydro-4H-benzo[b]pyrano[2,3-e][1,4]dioxin-4-one dihydrochloride pentahydrate

A3834.0005 A3834.0010
A3834.0005EA 2625 NOK
A3834.0005 A3834.0010
Spectinomycin dihydrochloride pentahydrate ≥95% (by titrimetric analysis), BioChemica®
Spectinomycin dihydrochloride pentahydrate
Formel: C₁₄H₂₄N₂O₇·2HCl·5H₂O
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00150886
CAS-nummer: 22189-32-8
EINECS: 244-554-3

Bestill nå

Spesifikasjon Testresultater

Assay (Titrimetric) Min. 95%

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR