Azure B (azure I) 89 Dye, certified_Biological Stain Commission, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Warning

Synonymer: C.I. 52010, N,N,N′-Trimethylthionin, Azure B, Trimethylthionine chloride, 3-(Dimethylamino)-7-(methylamino)phenothiazin-5-ium chloride, Methylene Azur B

MQ200

A4043-25G A4043-5G
A4043-25GEA 5260 NOK
A4043-25G A4043-5G
Azure B (azure I) 89 Dye, certified_Biological Stain Commission, Sigma-Aldrich®
Azure B (azure I)
Formel: C₁₅H₁₆ClN₃S
MW: 305.83 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011935
CAS-nummer: 531-55-5
EINECS: 208-511-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=600ef5cf3e062a4a587e204a7525059448b68bb0&vid=8472fabd8cb28ce018854ce4ca2ef091bb7e36ab&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR