qPCR Mycoplasma Test Kit
Leverandør: PanReac AppliChem
A9019.0025
A9019.0025EA 16675 NOK
A9019.0025
qPCR Mycoplasma Test Kit
Mikrobiologiske testsystemer Tester for mikroorganismer
Lyophilised PCR mix for the detection of mycoplasma in cell culture by Real-Time/qPCR.
Leveringsinformasjon: Supplied with Taq DNA polymerase. Kit contains PCR primer mix (incl. primers, probes, nucleotides), reaction buffer solution, water (PCR grade), positive template control, and internal control DNA.