Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10]

Leverandør: AAT BIOQUEST
10021000
AATB10021000EA 2925 NOK
AATB10021000
Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10]
Antistoffer
RPA-2.10 is an anti-human monoclonal antibody that is specific for the CD2 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

NB: For research use only.

Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: RPA-2.10
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR