Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (PE/iFluor® 700)

Leverandør: AAT BIOQUEST
100321X0 100321X1 100321X2
AATB100321X0EA 3750 NOK
AATB100321X0 AATB100321X1 AATB100321X2
Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (PE/iFluor® 700)
Antistoffer
The UCHT1 monoclonal antibody binds to human CD3e, a 20 kD member of the Ig superfamily commonly found on the surface of tregs, thymocytes, nkt cells and thymocytes (differentiation dependents).

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

NB: For research use only.

Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: UCHT1
Conjugation: PE/iFluor® 700
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol CD3e
Environmentally Preferable
Antigen-navn CD3e antigen
Antistofftype Primary
Klonalitet Monoclonal
Konjugering PE (Phycoerythrin)/iFluor® 700
Klon UCHT1
Reaktivitet Human
Vert Mouse
Isotype
ImmunoFluorescens Yes
Flowcytometri Yes
Rensing Affinity chromatography
Kyssabsorpsjon No
Lagringstemp 2...8 °C with minimised light exposure, do not freeze
Frakttemp. 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR