Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (PE/iFluor® 594)

Leverandør: AAT BIOQUEST
100321Y0 100321Y1 100321Y2
AATB100321Y0EA 3750 NOK
AATB100321Y0 AATB100321Y1 AATB100321Y2
Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (PE/iFluor® 594)
Antistoffer
The UCHT1 monoclonal antibody binds to human CD3e, a 20 kD member of the Ig superfamily commonly found on the surface of tregs, thymocytes, nkt cells and thymocytes (differentiation dependents).

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

NB: For research use only.

Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: UCHT1
Conjugation: PE/iFluor® 594
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR