Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (mFluor™ UV375)

Leverandør: AAT BIOQUEST
100330X0 100330X1
AATB100330X0EA 4350 NOK
AATB100330X0 AATB100330X1
Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (mFluor™ UV375)
Antistoffer
The SK7 monoclonal antibody reacts with human CD3e, a 20 kD member of the Ig superfamily often found on the surface of t cells, tregs and nkt cells.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

NB: For research use only.

Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: SK7
Conjugation: mFluor™ UV375
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR