Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (PB (Pacific Blue))

Leverandør: AAT BIOQUEST
100331J0 100331J1
AATB100331J0EA 4350 NOK
AATB100331J0 AATB100331J1
Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (PB (Pacific Blue))
Antistoffer
The SK7 monoclonal antibody reacts with human CD3e, a 20 kD member of the Ig superfamily often found on the surface of t cells, tregs and nkt cells.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

NB: For research use only.

Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: SK7
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR