Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (iFluor® 555)

Leverandør: AAT BIOQUEST
10041090 10041091
AATB10041090EA 4350 NOK
AATB10041090 AATB10041091
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (iFluor® 555)
Antistoffer
RPA-T4 is an anti-human monoclonal antibody that targets the CD4 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

NB: For research use only.

Type: Primary
Antigen: CD4 antigen
Clonality: Monoclonal
Clone: RPA-T4
Conjugation: iFluor® 555
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol CD4
Antigen-navn CD4 antigen
Environmentally Preferable
Antistofftype Primary
Klonalitet Monoclonal
Konjugering iFluor® 555
Klon RPA-T4
Reaktivitet Human
Vert Mouse
Isotype IgG1
ImmunoFluorescens Yes
Flowcytometri Yes
Rensing Affinity chromatography
Kyssabsorpsjon No
Lagringstemp 2...8 °C with minimised light exposure. Do not freeze.
Frakttemp. 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR