Anti-CD4 Mouse Monoclonal Antibody [clone: OKT-4] (iFluor® 594)

Leverandør: AAT BIOQUEST
100430C0
AATB100430C0EA 4350 NOK
AATB100430C0
Anti-CD4 Mouse Monoclonal Antibody [clone: OKT-4] (iFluor® 594)
Antistoffer
OKT-4 is an anti-human monoclonal antibody that targets the CD4 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

NB: For research use only.

Type: Primary
Antigen: CD4 antigen
Clonality: Monoclonal
Clone: OKT-4
Conjugation: iFluor® 594
Epitope:
Host: Mouse
Isotype: Mouse igg2b, κ
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR