Anti-CD5 Mouse Monoclonal Antibody [clone: L17F12] (iFluor® 488)

Leverandør: AAT BIOQUEST
10051050 10051051
AATB10051050EA 4350 NOK
AATB10051050 AATB10051051
Anti-CD5 Mouse Monoclonal Antibody [clone: L17F12] (iFluor® 488)
Antistoffer
L17F12 is an anti-human monoclonal antibody that is specific for the CD5 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

NB: For research use only.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: L17F12
Conjugation: iFluor® 488
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR