Anti-CD5 Mouse Monoclonal Antibody [clone: L17F12] (iFluor® 680)

Leverandør: AAT BIOQUEST
100510I0 100510I1
AATB100510I0EA 4350 NOK
AATB100510I0 AATB100510I1
Anti-CD5 Mouse Monoclonal Antibody [clone: L17F12] (iFluor® 680)
Antistoffer
L17F12 is an anti-human monoclonal antibody that is specific for the CD5 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

NB: For research use only.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: L17F12
Conjugation: iFluor® 680
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol CD5 CD5
Antigen-navn CD5 antigen CD5 antigen
Environmentally Preferable
Antistofftype Primary Primary
Klonalitet Monoclonal Monoclonal
Konjugering iFluor® 680 iFluor® 680
Klon L17F12 L17F12
Reaktivitet Human
Vert Mouse Mouse
Isotype
ImmunoFluorescens Yes Yes
Flowcytometri Yes Yes
Rensing Affinity chromatography Affinity chromatography
Kyssabsorpsjon No No
Lagringstemp 2...8 °C with minimized light exposure. Do not freeze. 2...8 °C with minimized light exposure. Do not freeze.
Frakttemp. 20 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR