Anti-CD5 Mouse Monoclonal Antibody [clone: L17F12] (APC)

Leverandør: AAT BIOQUEST
100511C0 100511C1 100511C2
AATB100511C0EA 3175 NOK
AATB100511C0 AATB100511C1 AATB100511C2
Anti-CD5 Mouse Monoclonal Antibody [clone: L17F12] (APC)
Antistoffer
L17F12 is an anti-human monoclonal antibody that is specific for the CD5 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

NB: For research use only.

Type: Primary
Antigen: CD5 antigen
Clonality: Monoclonal
Clone: L17F12
Conjugation: APC
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol CD5
Antigen-navn CD5 antigen
Environmentally Preferable
Antistofftype Primary
Klonalitet Monoclonal
Konjugering APC (Allophycocyanin)
Klon L17F12
Reaktivitet Human
Vert Mouse
Isotype
ImmunoFluorescens Yes
Flowcytometri Yes
Rensing Affinity chromatography
Kyssabsorpsjon No
Lagringstemp 2...8 °C with minimised light exposure. Do not freeze.
Frakttemp. 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR