Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (PacBlue)

Leverandør: AAT BIOQUEST
100811J0 100811J1
AATB100811J0EA 4350 NOK
AATB100811J0 AATB100811J1
Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (PacBlue)
Antistoffer
SK1 is an anti-human monoclonal antibody that is specific for the CD8a antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

NB: For research use only.

Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: SK1
Conjugation: PacBlue
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR