Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (PE/iFluor® 647)

Leverandør: AAT BIOQUEST
100811Q0 100811Q1 100811Q2
AATB100811Q0EA 3750 NOK
AATB100811Q0 AATB100811Q1 AATB100811Q2
Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (PE/iFluor® 647)
Antistoffer
SK1 is an anti-human monoclonal antibody that is specific for the CD8a antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

NB: For research use only.

Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: SK1
Conjugation: PE/iFluor® 647
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol CD8a
Environmentally Preferable
Antigen-navn CD8a antigen
Antistofftype Primary
Klonalitet Monoclonal
Konjugering PE (Phycoerythrin)/iFluor® 647
Klon SK1
Reaktivitet Human
Vert Mouse
Isotype
ImmunoFluorescens Yes
Flowcytometri Yes
Rensing Affinity chromatography
Kyssabsorpsjon No
Lagringstemp 2...8 °C with minimised light exposure, do not freeze
Frakttemp. 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR