Anti-CD14 Mouse Monoclonal Antibody [clone: 61D3] (iFluor® 660)

Leverandør: AAT BIOQUEST
101410G0 101410G1
AATB101410G0EA 4350 NOK
AATB101410G0 AATB101410G1
Anti-CD14 Mouse Monoclonal Antibody [clone: 61D3] (iFluor® 660)
Antistoffer
61D3 is an anti-human monoclonal antibody that targets the CD14 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

NB: For research use only.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: 61D3
Conjugation: iFluor® 660
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR