Anti-CD14 Mouse Monoclonal Antibody [clone: 61D3] (iFluor® 660)

Leverandør: AAT BIOQUEST
101410G0 101410G1
AATB101410G0EA 4350 NOK
AATB101410G0 AATB101410G1
Anti-CD14 Mouse Monoclonal Antibody [clone: 61D3] (iFluor® 660)
Antistoffer
61D3 is an anti-human monoclonal antibody that targets the CD14 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

NB: For research use only.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: 61D3
Conjugation: iFluor® 660
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol CD14
Environmentally Preferable
Antigen-navn CD14 antigen
Antistofftype Primary
Klonalitet Monoclonal
Konjugering iFluor® 660
Klon 61D3
Reaktivitet Human
Vert Mouse
Isotype
ImmunoFluorescens Yes
Flowcytometri Yes
Rensing Affinity chromatography
Kyssabsorpsjon No
Lagringstemp 2...8 °C with minimized light exposure. Do not freeze.
Frakttemp. 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR