Anti-CD16a Mouse Monoclonal Antibody [clone: 3G8] (PE/Texas Red®)

Leverandør: AAT BIOQUEST
101611S0 101611S1 101611S2
AATB101611S0EA 3750 NOK
AATB101611S0 AATB101611S1 AATB101611S2
Anti-CD16a Mouse Monoclonal Antibody [clone: 3G8] (PE/Texas Red®)
Antistoffer
The 3G8 monoclonal antibody recognises human CD16a, a 50 to 65 kD transmembrane protein typically found on the surface of macrophages, natural killer cells, dendritic cells, monocytes and neutrophils.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

NB: For research use only.

Type: Primary
Antigen: CD16a
Clonality: Monoclonal
Clone: 3G8
Conjugation: PE/Texas Red®
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR