Anti-CD16a Mouse Monoclonal Antibody [clone: 3G8] (PE/iFluor® 594)

Leverandør: AAT BIOQUEST
101611Y0 101611Y1 101611Y2
AATB101611Y0EA 3750 NOK
AATB101611Y0 AATB101611Y1 AATB101611Y2
Anti-CD16a Mouse Monoclonal Antibody [clone: 3G8] (PE/iFluor® 594)
Antistoffer
The 3G8 monoclonal antibody recognises human CD16a, a 50 to 65 kD transmembrane protein typically found on the surface of macrophages, natural killer cells, dendritic cells, monocytes and neutrophils.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

NB: For research use only.

Type: Primary
Antigen: CD16a
Clonality: Monoclonal
Clone: 3G8
Conjugation: PE/iFluor® 594
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol CD16a CD16a
Environmentally Preferable
Antigen-navn CD16a antigen CD16a antigen
Antistofftype Primary Primary
Klonalitet Monoclonal Monoclonal
Konjugering PE (Phycoerythrin)/iFluor® 594 PE (Phycoerythrin)/iFluor® 594
Klon 3G8 3G8
Reaktivitet Human
Vert Mouse Mouse
Isotype
ImmunoFluorescens Yes Yes
Flowcytometri Yes Yes
Rensing Affinity chromatography Affinity chromatography
Kyssabsorpsjon No No
Lagringstemp 2...8 °C with minimised light exposure, do not freeze 2...8 °C with minimised light exposure, do not freeze
Frakttemp. 20 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR