Anti-CD19 Mouse Monoclonal Antibody [clone: SJ25C1] (iFluor® 430)

Leverandør: AAT BIOQUEST
10191030 10191031
AATB10191030EA 4350 NOK
AATB10191030 AATB10191031
Anti-CD19 Mouse Monoclonal Antibody [clone: SJ25C1] (iFluor® 430)
Antistoffer
SJ25C1 is an anti-human monoclonal antibody that forms an immune complex with the CD19 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

NB: For research use only.

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: SJ25C1
Conjugation: iFluor® 430
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol CD19 CD19
Antigen-navn CD19 antigen CD19 antigen
Environmentally Preferable
Antistofftype Primary Primary
Klonalitet Monoclonal Monoclonal
Konjugering iFluor® 430 iFluor® 430
Klon SJ25C1 SJ25C1
Reaktivitet Human
Vert Mouse Mouse
Isotype
ImmunoFluorescens Yes Yes
Flowcytometri Yes Yes
Rensing Affinity chromatography Affinity chromatography
Kyssabsorpsjon No No
Lagringstemp 2...8 °C with minimized light exposure. Do not freeze. 2...8 °C with minimized light exposure. Do not freeze.
Frakttemp. 20 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR