Anti-CD19 Mouse Monoclonal Antibody [clone: SJ25C1] (FITC (Fluorescein Isothiocyanate))

Leverandør: AAT BIOQUEST
101911H0 101911H1
AATB101911H0EA 4350 NOK
AATB101911H0 AATB101911H1
Anti-CD19 Mouse Monoclonal Antibody [clone: SJ25C1] (FITC (Fluorescein Isothiocyanate))
Antistoffer
SJ25C1 is an anti-human monoclonal antibody that forms an immune complex with the CD19 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

NB: For research use only.

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: SJ25C1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR