iFluor® 405, Succinimidyl Ester
Leverandør: AAT BIOQUEST
1021 71021 71501 71551
AATB1021EA 3600 NOK
AATB1021 AATB71021 AATB71501 AATB71551
iFluor® 405, Succinimidyl Ester
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright Fluorescence
  • High Photostability
  • Low Quenching On Proteins
  • pH-Independent Signal
NB: For research use only.