iFluor® 532, Succinimidyl Ester
Leverandør: AAT BIOQUEST
1025 71025 71505 71555
AATB1025EA 3600 NOK
AATB1025 AATB71025 AATB71505 AATB71555
iFluor® 532, Succinimidyl Ester
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright Fluorescence
  • High Photostability
  • Low Quenching On Proteins
  • pH-Independent Signal
NB: For research use only.