Anti-CD27 Mouse Monoclonal Antibody [clone: O323] (iFluor® 647)

Leverandør: AAT BIOQUEST
102710F0
AATB102710F0EA 4350 NOK
AATB102710F0
Anti-CD27 Mouse Monoclonal Antibody [clone: O323] (iFluor® 647)
Antistoffer
O323 is an anti-human monoclonal antibody that is specific for the CD27 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

NB: For research use only.

Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone: O323
Conjugation: iFluor® 647
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR