Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (iFluor® 350)

Leverandør: AAT BIOQUEST
10280010 10280011
AATB10280010EA 4350 NOK
AATB10280010 AATB10280011
Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (iFluor® 350)
Antistoffer
CD28.2 is an anti-human monoclonal antibody that forms an immune complex with the CD28 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

NB: For research use only.

Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: CD28.2
Conjugation: iFluor® 350
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol CD28
Antigen-navn CD28 antigen
Environmentally Preferable
Antistofftype Primary
Klonalitet Monoclonal
Konjugering iFluor® 350
Klon CD28.2
Reaktivitet Human
Vert Mouse
Isotype
ImmunoFluorescens Yes
Flowcytometri Yes
Rensing Affinity chromatography
Kyssabsorpsjon No
Lagringstemp 2...8 °C with minimised light exposure, do not freeze
Frakttemp. 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR