Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (iFluor® 700)

Leverandør: AAT BIOQUEST
103400J0 103400J1
AATB103400J0EA 4350 NOK
AATB103400J0 AATB103400J1
Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (iFluor® 700)
Antistoffer
4H11 is an anti-human monoclonal antibody that recognises the CD34 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

NB: For research use only.

Type: Primary
Antigen: CD34 antigen
Clonality: Monoclonal
Clone: 4H11
Conjugation: iFluor® 700
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol CD34
Antigen-navn CD34 antigen
Environmentally Preferable
Antistofftype Primary
Klonalitet Monoclonal
Konjugering iFluor® 700
Klon 4H11
Reaktivitet Human
Vert Mouse
Isotype IgG1
ImmunoFluorescens Yes
Flowcytometri Yes
Rensing Affinity chromatography
Kyssabsorpsjon No
Lagringstemp 2...8 °C with minimised light exposure. Do not freeze.
Frakttemp. 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR