Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (iFluor® 840)

Leverandør: AAT BIOQUEST
103400Q0 103400Q1
AATB103400Q0EA 4350 NOK
AATB103400Q0 AATB103400Q1
Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (iFluor® 840)
Antistoffer
4H11 is an anti-human monoclonal antibody that recognises the CD34 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

NB: For research use only.

Type: Primary
Antigen: CD34 antigen
Clonality: Monoclonal
Clone: 4H11
Conjugation: iFluor® 840
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR