Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (iFluor® 532)

Leverandør: AAT BIOQUEST
10380070 10380071
AATB10380070EA 4350 NOK
AATB10380070 AATB10380071
Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (iFluor® 532)
Antistoffer
HIT2 is an anti-human monoclonal antibody that targets the CD38 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

NB: For research use only.

Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: HIT2
Conjugation: iFluor® 532
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol CD38
Environmentally Preferable
Antigen-navn CD38 antigen
Antistofftype Primary
Klonalitet Monoclonal
Konjugering iFluor® 532
Klon HIT2
Reaktivitet Human
Vert Mouse
Isotype IgG1
ImmunoFluorescens Yes
Flowcytometri Yes
Rensing Affinity chromatography
Kyssabsorpsjon No
Lagringstemp 2...8 °C with minimized light exposure. Do not freeze.
Frakttemp. 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR