Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (iFluor® 700)

Leverandør: AAT BIOQUEST
103800J0 103800J1
AATB103800J0EA 4350 NOK
AATB103800J0 AATB103800J1
Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (iFluor® 700)
Antistoffer
HIT2 is an anti-human monoclonal antibody that targets the CD38 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

NB: For research use only.

Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: HIT2
Conjugation: iFluor® 700
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR