Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (APC)

Leverandør: AAT BIOQUEST
103801C0 103801C1 103801C2
AATB103801C0EA 3175 NOK
AATB103801C0 AATB103801C1 AATB103801C2
Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (APC)
Antistoffer
HIT2 is an anti-human monoclonal antibody that targets the CD38 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

NB: For research use only.

Type: Primary
Antigen: CD38 antigen
Clonality: Monoclonal
Clone: HIT2
Conjugation: APC
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR